siRNA Details
| Gene Symbol: Zfp324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758845.1 | XP_008757067.1 | 2 | 1935 | ||
| XM_008758846.1 | XP_008757068.1 | 2 | 1909 | |||
| XM_008758847.1 | XP_008757069.1 | 2 | 2262 | |||
| XM_344862.7 | XP_344863.6 | 4 | 1651 | |||

Zfp324
Genomic Map