siRNA Details
| Gene Symbol: Micb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017468.2 | NP_001017468.1 | 1 | 65 | ||
| GenBank mRNA | AB113963.1 | - | 1 | 55 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Micb| Gene Symbol: Micb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017468.2 | NP_001017468.1 | 1 | 65 | ||
| GenBank mRNA | AB113963.1 | - | 1 | 55 | ||