siRNA Details
| Gene Symbol: Zdhhc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039037.1 | NP_001034126.1 | 4 | 386 | ||
| XM_006229258.2 | XP_006229320.1 | 4 | 405 | |||
| XM_008759402.1 | XP_008757624.1 | 4 | 384 | |||
| GenBank mRNA | AY886529.1 | - | 4 | 386 | ||

Zdhhc13
Genomic Map