siRNA Details
| Gene Symbol: Tmprss11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004020.1 | NP_001004020.1 | 3 | 202 | ||
| XM_008770049.1 | XP_008768271.1 | 3 | 296 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmprss11b| Gene Symbol: Tmprss11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004020.1 | NP_001004020.1 | 3 | 202 | ||
| XM_008770049.1 | XP_008768271.1 | 3 | 296 | |||