siRNA Details
| Gene Symbol: Rrm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013236.1 | NP_001013254.1 | 18 | 2266 | ||
| GenBank mRNA | BC085906.1 | - | 18 | 2266 | 7193834 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rrm1| Gene Symbol: Rrm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013236.1 | NP_001013254.1 | 18 | 2266 | ||
| GenBank mRNA | BC085906.1 | - | 18 | 2266 | 7193834 | |