siRNA Details
| Gene Symbol: Cyb5r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014244.1 | NP_001014266.1 | 9 | 813 | ||
| XM_008759774.1 | XP_008757996.1 | 7 | 2063 | |||
| XM_008759775.1 | XP_008757997.1 | 8 | 1966 | |||
| XM_008759776.1 | XP_008757998.1 | 8 | 1978 | |||
| XM_008759777.1 | XP_008757999.1 | 7 | 1951 | |||
| XM_008759778.1 | XP_008758000.1 | 8 | 1381 | |||
| XM_008759779.1 | XP_008758001.1 | 8 | 1538 | |||
| XM_008759780.1 | XP_008758002.1 | 6 | 567 | |||
| XM_008759781.1 | XP_008758003.1 | 9 | 862 | |||
| XM_008759782.1 | XP_008758004.1 | 7 | 1999 | |||
| GenBank mRNA | BC079235.1 | - | 9 | 813 | 7132617 | |

Cyb5r2
Genomic Map