siRNA Details
| Gene Symbol: Rras2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013434.1 | NP_001013452.1 | 5 | 709 | ||
| GenBank mRNA | BC091333.1 | - | 5 | 709 | 7314509 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rras2| Gene Symbol: Rras2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013434.1 | NP_001013452.1 | 5 | 709 | ||
| GenBank mRNA | BC091333.1 | - | 5 | 709 | 7314509 | |