siRNA Details
Gene Symbol: Tmc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012216.1 | NP_001012216.1 | 15, 16 | 2279 | ||
GenBank mRNA | BC088406.1 | - | 15, 16 | 2279 | 7112487 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012216.1 | NP_001012216.1 | 15, 16 | 2279 | ||
GenBank mRNA | BC088406.1 | - | 15, 16 | 2279 | 7112487 |