siRNA Details
| Gene Symbol: Stk32c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108922.1 | NP_001102392.1 | 4 | 701 | ||
| GenBank mRNA | FQ213397.1 | - | 3 | 302 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stk32c| Gene Symbol: Stk32c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108922.1 | NP_001102392.1 | 4 | 701 | ||
| GenBank mRNA | FQ213397.1 | - | 3 | 302 | ||