siRNA Details
| Gene Symbol: Clcf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207615.2 | NP_997498.2 | 1 | 320 | ||
| GenBank mRNA | BC098643.1 | - | 1 | 320 | 7123848 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clcf1| Gene Symbol: Clcf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207615.2 | NP_997498.2 | 1 | 320 | ||
| GenBank mRNA | BC098643.1 | - | 1 | 320 | 7123848 | |