siRNA Details
| Gene Symbol: Cpsf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014245.2 | NP_001014267.1 | 2 | 135 | ||
| XM_006231061.2 | XP_006231123.1 | 3 | 216 | |||
| XM_006231062.2 | XP_006231124.1 | 3 | 278 | |||
| XM_008760238.1 | XP_008758460.1 | 3 | 209 | |||
| XM_008760239.1 | XP_008758461.1 | 3 | 209 | |||
| GenBank mRNA | BC083864.1 | - | 2 | 138 | 7127736 | |

Cpsf7
Genomic Map