siRNA Details
Gene Symbol: Fra10ac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014246.1 | NP_001014268.1 | 4 | 327 | ||
XM_008760371.1 | XP_008758593.1 | 4 | 1316 | |||
XM_008760372.1 | XP_008758594.1 | 4 | 1316 | |||
GenBank mRNA | BC090036.1 | - | 4 | 327 | 7368097 |
