siRNA Details
| Gene Symbol: Lcor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006231418.2 | XP_006231480.1 | 2 | 3374 | ||
| XM_008760470.1 | XP_008758692.1 | 1 | 3351 | |||
| XM_008760471.1 | XP_008758693.1 | 1 | 3354 | |||
| XM_008760472.1 | XP_008758694.1 | 1 | 3350 | |||

Lcor
Genomic Map