siRNA Details
Gene Symbol: Cpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053526.2 | NP_445978.1 | 7 | 1236 | ||
GenBank mRNA | AB042599.1 | - | 7 | 1245 | ||
BC088124.1 | - | 7 | 1236 | 7326031 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053526.2 | NP_445978.1 | 7 | 1236 | ||
GenBank mRNA | AB042599.1 | - | 7 | 1245 | ||
BC088124.1 | - | 7 | 1236 | 7326031 |