siRNA Details
Gene Symbol: Cpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053526.2 | NP_445978.1 | 8, 9 | 1428 | ||
GenBank mRNA | AB042599.1 | - | 8, 9 | 1437 | ||
BC088124.1 | - | 8, 9 | 1428 | 7326031 | ||
FQ218535.1 | - | 3, 4 | 242 | |||
FQ219519.1 | - | 6, 7 | 713 |
