siRNA Details
| Gene Symbol: Cacul1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014248.1 | NP_001014270.1 | 5 | 996 | ||
| XM_006231678.2 | XP_006231740.1 | 5 | 1156 | |||
| GenBank mRNA | BC083839.1 | - | 5 | 996 | 7114271 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cacul1| Gene Symbol: Cacul1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014248.1 | NP_001014270.1 | 5 | 996 | ||
| XM_006231678.2 | XP_006231740.1 | 5 | 1156 | |||
| GenBank mRNA | BC083839.1 | - | 5 | 996 | 7114271 | |