siRNA Details
| Gene Symbol: RGD1563815 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127556.1 | NP_001121028.1 | 2 | 199 | ||
| XM_006256320.2 | XP_006256382.1 | 3 | 293 | |||
| XM_008772898.1 | XP_008771120.1 | 2 | 221 | |||
| GenBank mRNA | BC161883.1 | - | 2 | 55 | 7131276 | |
| Gene Symbol: LOC103694884 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773855.1 | XP_008772077.1 | 1 | 16 | ||

RGD1563815
Genomic Map