siRNA Details
| Gene Symbol: Rtdr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127557.2 | NP_001121029.1 | 2, 3 | 352 | ||
| XM_006256340.2 | XP_006256402.1 | 3, 4 | 408 | |||
| XM_006256341.2 | XP_006256403.1 | 3, 4 | 461 | |||
| GenBank mRNA | BC162057.1 | - | 2, 3 | 354 | 7114691 | |

Rtdr1
Genomic Map