siRNA Details
Gene Symbol: Pdss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014249.1 | NP_001014271.1 | 8 | 1304 | ||
GenBank mRNA | BC085898.1 | - | 8 | 1304 | 7193369 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pdss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014249.1 | NP_001014271.1 | 8 | 1304 | ||
GenBank mRNA | BC085898.1 | - | 8 | 1304 | 7193369 |