siRNA Details
Gene Symbol: Pdss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014249.1 | NP_001014271.1 | 2 | 614 | ||
XM_008773004.1 | XP_008771226.1 | 2 | 686 | |||
GenBank mRNA | BC085898.1 | - | 2 | 614 | 7193369 |
