siRNA Details
| Gene Symbol: Atg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014250.1 | NP_001014272.1 | 2 | 293 | ||
| GenBank mRNA | AM087012.1 | - | 2 | 293 | ||
| BC083602.1 | - | 2 | 293 | 7133143 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atg5| Gene Symbol: Atg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014250.1 | NP_001014272.1 | 2 | 293 | ||
| GenBank mRNA | AM087012.1 | - | 2 | 293 | ||
| BC083602.1 | - | 2 | 293 | 7133143 | ||