siRNA Details
| Gene Symbol: LOC365778 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014251.1 | NP_001014273.1 | 1 | 488 | ||
| GenBank mRNA | AY237129.1 | - | 340 | |||
| BC078989.1 | - | 1 | 488 | 7112531 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC365778| Gene Symbol: LOC365778 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014251.1 | NP_001014273.1 | 1 | 488 | ||
| GenBank mRNA | AY237129.1 | - | 340 | |||
| BC078989.1 | - | 1 | 488 | 7112531 | ||