siRNA Details
Gene Symbol: Selt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014253.2 | NP_001014275.2 | 4 | 502 | ||
GenBank mRNA | AY995234.1 | - | 4 | 502 | ||
BC086953.1 | - | 4 | 396 | 7321890 | ||
FQ211813.1 | - | 520 |
