siRNA Details
| Gene Symbol: Clk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014254.1 | NP_001014276.1 | 3 | 527 | ||
| XM_008761185.1 | XP_008759407.1 | 3 | 732 | |||
| XM_008761186.1 | XP_008759408.1 | 4 | 599 | |||
| XM_008761187.1 | XP_008759409.1 | 3 | 732 | |||
| XM_008761188.1 | XP_008759410.1 | 3 | 711 | |||
| XM_008761189.1 | XP_008759411.1 | 3 | 711 | |||
| XM_008761190.1 | XP_008759412.1 | 3 | 708 | |||
| XM_008761191.1 | XP_008759413.1 | 3 | 708 | |||
| XM_008761192.1 | XP_008759414.1 | 3 | 578 | |||
| XM_008761193.1 | XP_008759415.1 | 1 | 34 | |||
| GenBank mRNA | BC083788.1 | - | 3 | 527 | 7098251 | |

Clk2
Genomic Map