siRNA Details
| Gene Symbol: Clk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014254.1 | NP_001014276.1 | 6 | 894 | ||
| XM_008761185.1 | XP_008759407.1 | 6 | 1099 | |||
| XM_008761186.1 | XP_008759408.1 | 7 | 966 | |||
| XM_008761187.1 | XP_008759409.1 | 6 | 1096 | |||
| XM_008761188.1 | XP_008759410.1 | 6 | 1078 | |||
| XM_008761189.1 | XP_008759411.1 | 6 | 1075 | |||
| XM_008761190.1 | XP_008759412.1 | 6 | 1075 | |||
| XM_008761191.1 | XP_008759413.1 | 6 | 1072 | |||
| XM_008761192.1 | XP_008759414.1 | 6 | 945 | |||
| XM_008761193.1 | XP_008759415.1 | 3 | 746 | |||
| XM_008761194.1 | XP_008759416.1 | 3 | 225 | |||
| GenBank mRNA | BC083788.1 | - | 6 | 894 | 7098251 | |

Clk2
Genomic Map