siRNA Details
| Gene Symbol: Pip5k1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042621.1 | NP_001036086.1 | 6 | 846 | ||
| XM_008761315.1 | XP_008759537.1 | 6 | 975 | |||
| XM_008761316.1 | XP_008759538.1 | 6 | 972 | |||
| GenBank mRNA | AY724476.1 | - | 6 | 846 | ||

Pip5k1a
Genomic Map