siRNA Details
| Gene Symbol: Bola1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001071776.1 | NP_001065244.1 | 2 | 167 | ||
| GenBank mRNA | DQ912022.1 | - | 2 | 167 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bola1| Gene Symbol: Bola1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001071776.1 | NP_001065244.1 | 2 | 167 | ||
| GenBank mRNA | DQ912022.1 | - | 2 | 167 | ||