siRNA Details
| Gene Symbol: Alx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007012.1 | NP_001007013.1 | 3 | 712 | ||
| GenBank mRNA | AY488087.1 | - | 3 | 712 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alx3| Gene Symbol: Alx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007012.1 | NP_001007013.1 | 3 | 712 | ||
| GenBank mRNA | AY488087.1 | - | 3 | 712 | ||