siRNA Details
| Gene Symbol: Tmem56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135879.1 | NP_001129351.1 | 4 | 383 | ||
| XM_006233244.2 | XP_006233306.1 | 6 | 657 | |||
| XM_006233246.2 | XP_006233308.1 | 6 | 660 | |||
| XM_006233251.2 | XP_006233313.1 | 6 | 480 | |||
| XM_006233252.2 | XP_006233314.1 | 5 | 428 | |||
| XM_006233253.2 | XP_006233315.1 | 5 | 562 | |||
| XM_006233254.2 | XP_006233316.1 | 5 | 565 | |||
| XM_008761467.1 | XP_008759689.1 | 5 | 827 | |||
| XM_008761468.1 | XP_008759690.1 | 5 | 461 | |||

Tmem56
Genomic Map