siRNA Details
| Gene Symbol: Lhx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012219.1 | NP_001012219.1 | 6 | 833 | ||
| XM_006233528.2 | XP_006233590.1 | 7 | 1099 | |||
| GenBank mRNA | AF220000.1 | - | 1 | 93 | ||
| BC090011.1 | - | 6 | 833 | 7324565 | ||

Lhx8
Genomic Map