siRNA Details
| Gene Symbol: Lhx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012219.1 | NP_001012219.1 | 7 | 962 | ||
| XM_006233528.2 | XP_006233590.1 | 8 | 1228 | |||
| GenBank mRNA | BC090011.1 | - | 7 | 962 | 7324565 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lhx8| Gene Symbol: Lhx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012219.1 | NP_001012219.1 | 7 | 962 | ||
| XM_006233528.2 | XP_006233590.1 | 8 | 1228 | |||
| GenBank mRNA | BC090011.1 | - | 7 | 962 | 7324565 | |