siRNA Details
| Gene Symbol: Nup188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006233905.2 | XP_006233967.1 | 10, 11 | 972 | ||
| XM_006233906.2 | XP_006233968.1 | 10, 11 | 1462 | |||
| XM_345335.7 | XP_345336.2 | 10, 11 | 972 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nup188| Gene Symbol: Nup188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006233905.2 | XP_006233967.1 | 10, 11 | 972 | ||
| XM_006233906.2 | XP_006233968.1 | 10, 11 | 1462 | |||
| XM_345335.7 | XP_345336.2 | 10, 11 | 972 | |||