siRNA Details
| Gene Symbol: Wdr38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761808.1 | XP_008760030.1 | 8 | 934 | ||
| XM_575125.5 | XP_575125.6 | 8 | 894 | |||
| GenBank mRNA | FQ213095.1 | - | 8 | 888 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr38| Gene Symbol: Wdr38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761808.1 | XP_008760030.1 | 8 | 934 | ||
| XM_575125.5 | XP_575125.6 | 8 | 894 | |||
| GenBank mRNA | FQ213095.1 | - | 8 | 888 | ||