siRNA Details
| Gene Symbol: RGD1564319 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126290.1 | NP_001119762.1 | 1 | 274 | ||
| GenBank mRNA | BC160905.1 | - | 1 | 274 | 7455398 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1564319| Gene Symbol: RGD1564319 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126290.1 | NP_001119762.1 | 1 | 274 | ||
| GenBank mRNA | BC160905.1 | - | 1 | 274 | 7455398 | |