siRNA Details
| Gene Symbol: Esf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100771.1 | NP_001094241.1 | 6 | 1372 | ||
| XM_006235105.2 | XP_006235167.1 | 6 | 1505 | |||
| XM_006235106.2 | XP_006235168.1 | 6 | 1371 | |||
| GenBank mRNA | AB038233.1 | - | 6 | 1370 | ||
| BC105755.1 | - | 6 | 1306 | |||

Esf1
Genomic Map