siRNA Details
| Gene Symbol: Nkx2-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_345445.7 | XP_345446.3 | 2 | 950 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nkx2-4| Gene Symbol: Nkx2-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_345445.7 | XP_345446.3 | 2 | 950 | ||