siRNA Details
| Gene Symbol: Apmap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034003.1 | NP_001029175.1 | 8 | 1022 | ||
| XM_006235215.2 | XP_006235277.1 | 9 | 1156 | |||
| XM_006235217.2 | XP_006235279.1 | 9 | 1039 | |||
| XM_008762362.1 | XP_008760584.1 | 9 | 1570 | |||
| GenBank mRNA | BC090021.1 | - | 4 | 506 | ||
| BC105824.1 | - | 8 | 1022 | 7372968 | ||

Apmap
Genomic Map