siRNA Details
| Gene Symbol: Apmap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034003.1 | NP_001029175.1 | 3 | 308 | ||
| XM_006235215.2 | XP_006235277.1 | 4 | 442 | |||
| XM_006235217.2 | XP_006235279.1 | 4 | 325 | |||
| XM_008762362.1 | XP_008760584.1 | 4 | 856 | |||
| GenBank mRNA | AY325201.1 | - | 5 | 346 | ||
| BC105824.1 | - | 3 | 308 | 7372968 | ||

Apmap
Genomic Map