siRNA Details
| Gene Symbol: R3hdml | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108962.1 | NP_001102432.1 | 3 | 573 | ||
| XM_006235600.2 | XP_006235662.1 | 4 | 716 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

R3hdml| Gene Symbol: R3hdml | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108962.1 | NP_001102432.1 | 3 | 573 | ||
| XM_006235600.2 | XP_006235662.1 | 4 | 716 | |||