siRNA Details
| Gene Symbol: Rbm38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108965.1 | NP_001102435.1 | 3 | 1329 | ||
| XM_006235704.2 | XP_006235766.1 | 4 | 799 | |||
| XM_008762528.1 | XP_008760750.1 | 5 | 889 | |||
| GenBank mRNA | FQ227741.1 | - | 1 | 116 | ||

Rbm38
Genomic Map