siRNA Details
| Gene Symbol: Polr3k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014259.1 | NP_001014281.1 | 2 | 245 | ||
| GenBank mRNA | BC089990.1 | - | 2 | 245 | 7319381 | |
| FQ212572.1 | - | 2 | 267 | |||
| FQ213710.1 | - | 2 | 268 | |||
| FQ213830.1 | - | 2 | 279 | |||
| FQ216547.1 | - | 2 | 268 | |||
| FQ226947.1 | - | 2 | 250 | |||
| FQ232144.1 | - | 2 | 279 | |||

Polr3k
Genomic Map