siRNA Details
Gene Symbol: Clvs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108969.1 | NP_001102439.1 | 4, 5 | 1126 | ||
GenBank mRNA | FQ214563.1 | - | 3, 4 | 951 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Clvs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108969.1 | NP_001102439.1 | 4, 5 | 1126 | ||
GenBank mRNA | FQ214563.1 | - | 3, 4 | 951 |