siRNA Details
| Gene Symbol: Igfbpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108972.2 | NP_001102442.1 | 2 | 679 | ||
| GenBank mRNA | BC158647.1 | - | 2 | 679 | 8364465 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Igfbpl1| Gene Symbol: Igfbpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108972.2 | NP_001102442.1 | 2 | 679 | ||
| GenBank mRNA | BC158647.1 | - | 2 | 679 | 8364465 | |