siRNA Details
| Gene Symbol: Oxct2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012221.1 | NP_001012221.1 | 1 | 1581 | ||
| GenBank mRNA | BC083681.1 | - | 1 | 1581 | 7135892 | |
| Gene Symbol: LOC100361036 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008764112.1 | XP_008762334.1 | 1 | 2131 | ||

Oxct2a
Genomic Map