siRNA Details
| Gene Symbol: Azin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC078981.1 | - | 1265 | 7112100 | ||
| HQ413774.1 | - | 8 | 1114 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Azin2| Gene Symbol: Azin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC078981.1 | - | 1265 | 7112100 | ||
| HQ413774.1 | - | 8 | 1114 | |||