siRNA Details
Gene Symbol: Wrap73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014262.1 | NP_001014284.1 | 2 | 197 | ||
GenBank mRNA | BC082062.1 | - | 2 | 197 | 7132552 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Wrap73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014262.1 | NP_001014284.1 | 2 | 197 | ||
GenBank mRNA | BC082062.1 | - | 2 | 197 | 7132552 |