siRNA Details
| Gene Symbol: Ttc39d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191906.1 | NP_001178835.1 | 2 | 1813 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttc39d| Gene Symbol: Ttc39d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191906.1 | NP_001178835.1 | 2 | 1813 | ||