siRNA Details
| Gene Symbol: Dnajc5g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013242.1 | NP_001013260.1 | 3, 4 | 376 | ||
| XM_006239826.2 | XP_006239888.1 | 3, 4 | 437 | |||
| XM_006239827.2 | XP_006239889.1 | 3, 4 | 437 | |||
| GenBank mRNA | BC083671.1 | - | 3, 4 | 376 | 7135668 | |

Dnajc5g
Genomic Map