siRNA Details
| Gene Symbol: Dus4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135803.1 | NP_001129275.1 | 5 | 561 | ||
| XM_006239990.2 | XP_006240052.1 | 5 | 492 | |||
| XM_006239991.1 | XP_006240053.1 | 6 | 719 | |||
| XM_006239992.2 | XP_006240054.1 | 6 | 996 | |||
| XM_006239993.2 | XP_006240055.1 | 4 | 405 | |||
| XM_008764601.1 | XP_008762823.1 | 4 | 367 | |||

Dus4l
Genomic Map