siRNA Details
| Gene Symbol: Cfl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108982.2 | NP_001102452.2 | 4 | 1545 | ||
| XM_006240104.2 | XP_006240166.1 | 4 | 1632 | |||
| GenBank mRNA | FQ224178.1 | - | 4 | 1221 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cfl2| Gene Symbol: Cfl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108982.2 | NP_001102452.2 | 4 | 1545 | ||
| XM_006240104.2 | XP_006240166.1 | 4 | 1632 | |||
| GenBank mRNA | FQ224178.1 | - | 4 | 1221 | ||